missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 038710.AP

Code : NEW

Additional Details : CAS Number : 1336-21-6 Weight : 0.01000kg

 View more versions of this product

Product Code. 11460161

  • £20.50 / 500mL
Estimated Shipment: 28-08-2024
to see stock.

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution
1336-21-6
H5NO
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
14923
Specpure™
pH 10.00 (±0.01 at 25°C), No Color, NIST Traceable
500 mL
MFCD00066650
N.O
35.05
CHEBI:18219
Liquid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only