missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, Reagecon™
BIOPHARMA

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 10.00 is tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

Brand:  Reagecon 1100

 View more versions of this product

Product Code. 11412024

  • £48.57 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
pH Buffer
Yes
100°C
10.0 at 20°C
1000 mL
Characteristic
Fully miscible in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 @ 20°C

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1024F1 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number1100
Lot Number1024F1 (1) Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number1100
Lot Number1023K1 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number1100
Lot Number1023K1 (2) Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number1100
Lot Number1023K1 (3) Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number1100
Special Offers

Special Offers

  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    12

    Minutes

    17

    Seconds

    59
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution, pH 10.00, Reagecon™ > Quantity: 1000mL; Tested at 20 deg C

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.