missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10, Mettler Toledo™
Shop All Mettler Toledo Products

Click to view available options
Quantity:
250 mL
Packaging:
1 Pk.
6 Pk.
Description
Greener Choice Claims
- Environmental benefits include:
- Certifications and Seals of Approval
- Renewable Energy

Specifications
Specifications
Color | Colorless |
pH | 10 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | pH Buffer |
Odor | None |
Packaging | 6 Pk. |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution, pH 10, Mettler Toledo™ >
Spot an opportunity for improvement?Share a Content Correction