missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 13.00, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Color | Colorless |
| pH | 13.00 ±0.05 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | pH Buffer |
| Odor | Characteristic |
| Traceability to NIST | Yes |
| Solubility Information | Fully miscible in water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction