missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Ph 2.0 (20 °C), Honeywell Fluka™
Citric acid / hydrochloric acid / sodium chloride, with fungicide, traceable to SRM from NIST and PTB
Brand: Honeywell-Fluka 31045-1L
34.98 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer Solution Ph 2.0 (20 °C) | |
7732-18-5, 7647-14-5, 77-92-9 | |
MFCD00146206 | |
1789 |
With fungicide | |
1 L | |
Plastic Bottle |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : citric acid, sodiumchloride, hydrochloric acid