Learn More
Buffer Solution Ph 2.0 (20 °C), Honeywell Fluka™
Citric acid / hydrochloric acid / sodium chloride, with fungicide, traceable to SRM from NIST and PTB
Brand: Honeywell Fluka 31045-6X1L
Code : NEW
Additional Details : CAS Number : 7647-14-5 Weight : 6.01800kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer Solution Ph 2.0 (20°C) | |
7732-18-5,7647-14-5,77-92-9 | |
MFCD00146206 |
With fungicide | |
6 x 1 L | |
UN1789 |
Safety and Handling
- Buffer solution pH 2.0 (20 °C)
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Citric acid, Sodiumchloride, hydrochloric acid