missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 2.00, Reagecon™
BIOPHARMA

Product Code. p-8015626 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 11492004

Brand: Reagecon 10205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 2.00 is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 2.00 ±0.02
Physical Form Liquid
Quantity 500 mL
Formulation Glycine, Hydrochloric Acid, Sodium Chloride
Traceability to NIST Yes
Solubility Information Fully miscible in water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.