missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™ Product Code.: 10578040

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™

Product Code. 10578040
1 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10578040

Brand: Fisher Chemical J/2800/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 217672

Specifications

Color Colorless
pH 2
Physical Form Liquid
Quantity 1 L
Packaging HDPE plastic bottle
compliance-icons
Product Identifier
  • Buffer solution pH 2 (glycine) Standard Buffer Solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.