missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 2, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
pH | 2 |
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution pH 2, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction