missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 3.00, Reagecon™
Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 3.00 is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.
£35.95 - £53.45
Specifications
Melting Point | 0°C |
---|---|
Boiling Point | 100°C |
Color | Colorless |
pH | 3.00 ±0.02 |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402014
|
Reagecon
10305 |
500 mL |
£35.95
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490449
|
Reagecon
1030 |
1 L |
£53.45
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
0°C | |
Colorless | |
Liquid | |
Characteristic | |
Fully miscible in water |
100°C | |
3.00 ±0.02 | |
pH Buffer | |
Yes |