missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 100°C |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000%, <1.0000% |
CAS | 18996-35-5, 144-33-2, 7647-14-5 |
Density | 1.0 g/cm3 |
Quantity | 6 x 500 mL |
Solution Type | Buffer |
Packaging | Poly Bottle |
Chemical Name or Material | Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'