Learn More
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable, Thermo Scientific Chemicals
£20.50 - £219.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490161
|
Thermo Scientific Alfa Aesar
038713.AP |
500 mL |
£20.50
500mL |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11378987
|
Thermo Scientific Alfa Aesar
038713.K2 |
1 L |
£32.50
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11388987
|
Thermo Scientific Alfa Aesar
038713.K7 |
4 L |
£59.70
4 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11400171
|
Thermo Scientific Alfa Aesar
038713.M4 |
20 L |
£219.00
20 L |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.