missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 383830010

35.13 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.


 View more versions of this product

Product Code. 10270023

  • £39.50 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer solution pH 4
1.0% (Potassium hydrogen phthalate)
1 L
MFCD00217672
204.22
877-24-7, 7732-18-5, 7487-94-7
Liquid
C8H5KO4
Solubility in water: miscible.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only