missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 4, Red, Reagecon™
BIOPHARMA

Product Code. 11424589 Shop All Reagecon Products
1 L, HDPE Twin Neck Bottle, 4 at 20°C
Click to view available options
Quantity:
250 mL
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle GL45
HDPE Twin Neck Bottle
pH:
4 at 20°C
4 at 25°C
This item is not returnable. View return policy
This item is not returnable. View return policy

Colored pH Buffer solutions in Twin-neck containers. Reagecon™ Buffer Solution pH 4, Red are ISO 17025 accredited and NIST traceable.

  • Colored
  • Twin-neck containers with integrated calibrating chamber
  • Tested at 20°C or 25°C
  • Certified by ISO 17025 Accredited Test Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Color Red
pH 4 at 20°C
Quantity 1 L
Packaging HDPE Twin Neck Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 4, Red, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.