missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 4, Red, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
250 mL
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle GL45
HDPE Twin Neck Bottle
pH:
4 at 20°C
4 at 25°C
Description
- Colored
- Twin-neck containers with integrated calibrating chamber
- Tested at 20°C or 25°C
- Certified by ISO 17025 Accredited Test Method
- NIST traceable
- Presented in various pack sizes
Specifications
Specifications
Color | Red |
pH | 4 at 25°C |
Quantity | 500 mL |
Packaging | HDPE Twin Neck Bottle |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4C24L1 | Certificate of Analysis | 11/02/2025 | 1040525CTT |
4C24H1 | Certificate of Analysis | 15/10/2024 | 1040525CTT |
4C24F1 | Certificate of Analysis | 07/08/2024 | 1040525CTT |
4C24E1 | Certificate of Analysis | 18/06/2024 | 1040525CTT |
4C24A2 | Certificate of Analysis | 22/04/2024 | 1040525CTT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution pH 4, Red, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction