missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution pH 5.00, Reagecon™ Product Code.: 11432014

Buffer Solution pH 5.00, Reagecon™

Product Code. 11432014 Shop All Reagecon Products
500 mL
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy

Product Code. 11432014

Brand: Reagecon 10505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH 5.00
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number524F1 Certificate TypeCertificate of Analysis Date01/02/2025 Catalog Number10505
Lot Number523M1 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number10505
Lot Number523C1 Certificate TypeCertificate of Analysis Date01/11/2023 Catalog Number10505
Lot Number522D1 Certificate TypeCertificate of Analysis Date23/03/2023 Catalog Number10505
Lot Number520J1 Certificate TypeCertificate of Analysis Date16/04/2022 Catalog Number10505
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 5.00, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.