missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 6.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 042418.K2

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11374578

  • £30.90 / 1 L
In Stock
to see stock.

Description

Description

Buffer solution (pH 6) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution, pH 6.00 (±0.01 at 25°C)
1 L
Specpure™
Liquid
NIST Traceable
MFCD00134130
Miscible with water
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only