missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution pH 6.8, Reagecon™ Product Code.: 11494549

Buffer Solution pH 6.8, Reagecon™

Product Code. 11494549 Shop All Reagecon Products
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11494549

Brand: Reagecon 106825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH 6.8
Quantity 1 L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number682524H1 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog Number106825
Lot Number682523A1 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number106825
Lot Number682521J1 Certificate TypeCertificate of Analysis Date06/07/2022 Catalog Number106825
Lot Number682521E1 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number106825
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 6.8, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.