missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 6.865, Reagecon™
BIOPHARMA

Product Code. 13116269
Click to view available options
Quantity:
500 mL
Packaging:
HDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for use where very accurate measurements are required. Reagecon pH Buffer Standard 6.865 is manufactured to exacting specifications with an extended shelf life.

A Professional Range buffer (per DIN 19266 value) that is supplied with a detailed Certificate of Analysis which outlines traceability to NIST.

  • Temperature dependence data, lot numbers and expiry dates printed on the label
  • Tested and ISO 17025 Certified

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
Density 1.0001g/cm3
pH 6.865
Physical Form Liquid
Quantity 500 mL
Packaging HDPE bottle
Traceability to NIST Traceable to NIST
Vapor Pressure 23hPa (at 20°C)
Solubility Information Fully miscible
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'