missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 6.865, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£39.40 - £57.55
Specifications
pH | 6.865 |
---|---|
Physical Form | Liquid |
Solution Type | Buffer |
Formulation | Phosphate Potassium, Dihydrogen/Phosphate Hydrogen |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12940654
|
Fisher Chemical
SOL 010 500 |
500 mL |
£39.40
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12900664
|
Fisher Chemical
SOL 016 1L |
1000 mL |
£57.55
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
6.865 | |
Liquid | |
Buffer | |
Phosphate Potassium, Dihydrogen/Phosphate Hydrogen |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title