missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™
31.05 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
6 x 100 mL
6 x 500 mL
10 L
6 x 1 L
Description
Leading the Industry For Over 65 Years. Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.
7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
Composition per liter:
3.52 g Potassium dihydrogen phosphate (KH2PO4)7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 7.0 |
Quantity | 1 L |
MDL Number | MFCD00066650 |
UN Number | NONH for all modes of transport |
IUPAC Name | azanium;hydroxide |
Chemical Name or Material | Buffer solution |
Safety and Handling
Product Identifier
- Buffer solution pH 7.0 (20 °C)
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'