missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
£19.80 - £57.00
Specifications
Chemical Name or Material | Buffer solution |
---|---|
CAS | 1336-21-6 |
Color | Colorless |
Physical Form | Liquid |
Molecular Formula | H5NO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480161
|
Thermo Scientific Alfa Aesar
038712.AP |
500 mL |
£19.80
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368987
|
Thermo Scientific Alfa Aesar
038712.K2 |
1 L |
£35.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358987
|
Thermo Scientific Alfa Aesar
038712.K7 |
4 L |
£57.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
H5NO | |
MFCD00066650 | |
pH 7.00 buffers are in solution form. Miscible with water. | |
N.O | |
14923 | |
Specpure™ |
1336-21-6 | |
Liquid | |
Odorless | |
Yes | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
35.05 | |
CHEBI:18219 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only