missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.00, Reagecon™
BIOPHARMA

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 7.00 is tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

Brand:  Reagecon 1070525

Product Code. 11476404

  • £36.61 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Calibrated to a standard precision NIST value
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
Characteristic
Fully miscible in water
100°C
7.00 ±0.01
500 mL
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number72525A1 Certificate TypeCertificate of Analysis Date15/04/2025 Catalog Number1070525
Lot Number72524K1 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number1070525
Lot Number72524G1 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog Number1070525
Lot Number72524C1 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog Number1070525
Lot Number72524K1 (1) Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number1070525
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution, pH 7.00, Reagecon™ > Quantity: 500mL; Tested at 25 deg C

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.