missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7, Baker Analyzed, Yellow, J.T. Baker™

Product Code. 15587614 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
Unit Size:
12 sheets
This item is not returnable. View return policy

Product Code. 15587614

Brand: Avantor J.T.Baker 565601

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Chemical Name or Material Buffer, pH 7 Reference standard solution
Color Yellow
Physical Form Liquid
Quantity 500 mL
Flash Point 0°C (32°F)
Packaging Poly
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.