Learn More
Buffer solution pH 7, Phosphate buffer, Thermo Scientific Chemicals
Marca: Thermo Scientific Acros 258591000
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Detalles adicionales : CAS Number : 7558-79-4 Weight : 0.15000kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferEspecificaciones
Buffer solution pH 7 | |
1.0030g/mL | |
H2KO4P | |
Plastic bottle | |
14, 7659 | |
Solubility in water: soluble. |
7558-79-4,7732-18-5 | |
100 mL | |
MFCD03097944 | |
1.003 | |
14mmHg at 20°C | |
136.09 |
RUO – Research Use Only