missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Coded pH Buffers in Twin Neck Bottle Format, Reagecon™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
250 mL
500 mL
1 L
Color:
Blue
Yellow
pH:
7
10
Description
- Colored pH Buffer Solutions
- Tested at 20°C or 25°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
- No possibility of contamination
- No need for separate measuring container for use in the calibration of the Electrode
- Correct quantity of buffer required for calibration is dispensed into the calibrating chamber giving rise to no waste
- Ideally suited for field work
- Easy to carry
- 250ml, 500ml and 1L sizes available
Specifications
Color | Blue |
pH | 10 |
Quantity | 250 mL |
Packaging | HDPE Twin Neck Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'