missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution, pH 8.00, Reagecon™ Product Code.: 11482014
compliance-icons

Buffer Solution, pH 8.00, Reagecon™
BIOPHARMA

Product Code. 11482014 Shop All Reagecon Products
500 mL, 8
Click to view available options
Quantity:
500 mL
1 L
pH:
8
This item is not returnable. View return policy

Product Code. 11482014

Brand: Reagecon 10805

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Certified by an ISO 17025 accredited test method and NIST traceable. Reagecon™ Buffer Solution, pH 8.00 is available in solutions tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 8
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Traceability to NIST Yes
Solubility Information Fully miscible in water
compliance-icons
Product Identifier
  • BUFFER SOLUTION pH 8.00 ± 0.01 @ 20°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Boric Acid, DisodiumSalt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution, pH 8.00, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.