missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 383870010
34.65 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution pH 9.2 | |
1330-43-4, 7732-18-5, 7647-14-5 | |
1 L | |
Plastic bottle | |
Solubility in water: miscible. |
Borate buffer, traceable to NIST, ready to use | |
1.0000g/mL | |
MFCD00163147 | |
1 |
Safety and Handling
Product Identifier
- Buffer solution pH 9.2, Borate buffer, traceable to NIST
RUO – Research Use Only