missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 9.21, METTLER TOLEDO™
Shop All Mettler Toledo Products

Click to view available options
Quantity:
250 mL
1000 mL
6 x 250 mL
Description
Greener Choice Claims
- Environmental benefits include:
- Certifications and Seals of Approval
- Renewable Energy

Specifications
Specifications
Color | Blue |
pH | 9.21 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | pH Buffer |
Odor | None |
Packaging | 250mL |
Safety and Handling

Signal Word
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution, pH 9.21, METTLER TOLEDO™ >
Spot an opportunity for improvement?Share a Content Correction