missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Product Code. p-8015115 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 10789234

Brand: Fisher Chemical J/2880/08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement

Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
  • Standard and color-coded buffer solutions
  • Manufactured to a tolerance of ±0.02 pH units
  • Accurately standardized and labelled with lot-specific pH at 20°C
  • All buffers are traceable to NIST and BS 1647 standard
  • Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
  • Standard buffers are available over the range pH2 to pH10
  • Color-coded buffer solutions are available over the range pH4, pH7, and pH
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 217672

Specifications

Color Colorless
pH 10
Physical Form Liquid
Quantity 500 mL
Packaging HDPE plastic bottle
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.