missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ Product Code.: 10111430

Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Product Code. 10111430
1 L, HDPE plastic bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy

Product Code. 10111430

Brand: Fisher Chemical J/2810/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

MDL Number 217672

Specifications

Color Colorless
pH 3
Physical Form Liquid
Quantity 1 L
Packaging HDPE plastic bottle
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    26

    Hours

    7

    Minutes

    33

    Seconds

    7
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.