Learn More
Buffer Solution pH4.00 Bs formulated, Standard Solution ready to use for pH measurement, Fisher Chemical
Brand: Fisher Chemical J/2821/15
92.09 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 23
Additional Details : CAS Number : 877-24-7 Weight : 1.00000kg
Specifications
Colorless | |
Liquid | |
282720 |
4 | |
1 L | |
Plastic container |