Learn More
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand:  Fisher Chemical J/2830/17
 View more versions of this product
Product Code. 10294430
  • £155.00 / 2.50 L
Edge
Explore more special offers
Description
Description
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement
TRUSTED_SUSTAINABILITY
Specifications
Specifications
877-24-7
5
2.5 L
HDPE plastic bottle
Colorless
Liquid
13070