Learn More
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2830/15
49.83 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 23
Additional Details : CAS Number : 877-24-7 Weight : 1.00000kg
Description
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement
Specifications
877-24-7 | |
5 | |
1 L | |
HDPE plastic bottle |
Colorless | |
Liquid | |
13070 |