Learn More
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2830/08
41.19 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 23
Additional Details : CAS Number : 877-24-7 Weight : 0.50000kg
Description
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement
Specifications
877-24-7 | |
5 | |
500 mL | |
HDPE plastic bottle |
Colorless | |
Liquid | |
13070 |