missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH6.87 BS formulated, Standard Solution ready to use for pH measurement, Fisher Chemical
Click to view available options
Quantity:
1 L
Packaging:
Plastic container
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
MDL Number | 282720 |
Packaging | Plastic container |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'