missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2860/08
27.79 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement
Specifications
Colorless | |
Liquid | |
217672 |
8 | |
500 mL | |
HDPE plastic bottle |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.