missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution SrCO3 for AAS 0.05mg/L HNO3 matrix, Fisher chemical™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Specifications
Concentration | 0.05mg/L |
Packaging | Bottle |
Quantity | 100 mL |
Type | Buffer Solution |
Solvent or Matrix | HNO3 |
Physical Form | Liquid |
Chemical Name or Material | SrCO3 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'