missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution SrCO3 for AAS 0.05mg/L HNO3 matrix, Fisher chemical™

Catalog Number 12944555
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 0.05mg/L
Packaging Bottle
Quantity 500 mL
Type Buffer Solution
Solvent or Matrix HNO3
Physical Form Liquid
Chemical Name or Material SrCO3
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'