missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffer Solutions for NucleoMag™ RNA
Additional buffer solutions (e.g., lysis buffer MR1, wash buffer MR3) for use in magnetic bead-based isolation of RNA from cell and tissue samples using the NucleoMag™ RNA kit.
Brand: Macherey-Nagel™ 744351.125
This item is not returnable.
View return policy
Description
NucleoMag™ RNA
- One-tube procedure minimizes risk of cross-contamination
- Proven 96-well plate format
- Small elution volumes ≥ 50 μL
- Suitable for manual and automated processing
- Recombinant DNase included
- Reducing agent TCEP included (no β-mercaptoethanol necessary)
Specifications
Colorless | |
Guanidinium thiocyanate; Water | |
125 mL | |
Odorless | |
Liquid | |
Lysis Buffer MR1 |
NucleoMag™ RNA kit | |
6.5 to 7.5 | |
593-84-0 | |
Room Temperature (18°C to 25°C) | |
Isolation of RNA from tissue and cell samples |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.