missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer Solutions for NucleoMag™ RNA

Additional buffer solutions (e.g., lysis buffer MR1, wash buffer MR3) for use in magnetic bead-based isolation of RNA from cell and tissue samples using the NucleoMag™ RNA kit.

Brand:  Macherey-Nagel™ 744353.500


 View more versions of this product

Product Code. 15889147

  • £88.20 / 320mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NucleoMag™ RNA

  • One-tube procedure minimizes risk of cross-contamination
  • Proven 96-well plate format
  • Small elution volumes ≥ 50 μL
  • Suitable for manual and automated processing
  • Recombinant DNase included
  • Reducing agent TCEP included (no β-mercaptoethanol necessary)
Specifications

Specifications

Colorless
Guanidinium thiocyanate; Ethanol; Water
593-84-0
Room Temperature (18°C to 25°C)
Isolation of RNA from tissue and cell samples
NucleoMag™ RNA kit
320 mL
Alcoholic
Liquid
Wash Buffer MR3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer MR3 (320 mL)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate, ethanol, Tergitol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers