Learn More
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Brand: Fisher Chemical B/4765/77
64.07 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 21
Additional Details : CAS Number : 7758-11-4 Weight : 0.05000kg
Description
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Specifications
White | |
Solid | |
Disodium Orthophosphate Heptahydrate, Sodium Chloride, Dipotassium Phosphate | |
Glass Jar |
7 | |
50 Tablets | |
81857 | |
Soluble |