Learn More
Thermo Scientific™ Buffered Peptone Water (Bottled)
Pre-enrich food samples prior to the isolation of Salmonella species using these ready-prepared media bottles.
£53.82 - £220.00
Specifications
Color | Straw, Clear |
---|---|
Content And Storage | 2 to 25°C |
Format | Sirop Bottle |
Manufacturing Origin | United Kingdom |
Product Type | Agar |
Product Code | Brand | Fill Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Fill Weight | Price | Quantity & Availability | |||||
13255469
|
Thermo Scientific™
BO0201S |
225.0g - 229.0g |
£220.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11962632
|
Thermo Scientific™
BO0201Z |
90g + 1.5g |
£53.82
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
Pre-enrich food samples prior to the isolation of Salmonella species with Thermo Scientific™ Oxoid™ Buffered Peptone Water ready-prepared bottled medium. Buffered Peptone Water is a non-selective pre-enrichment medium.
Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.
- Convenient: the pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
- Aids recovery of damaged Salmonella cells.
The test sample is added to Buffered Peptone Water at a ratio of 1:10 and incubated at 37° ± 1°C for 18 hours + 2 hours.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Straw, Clear | |
Sirop Bottle | |
Agar | |
10/Pk. |
2 to 25°C | |
United Kingdom | |
10 bottles with screw cap closures in a box |