Learn More
Thermo Scientific™ Buffered Peptone Water (ISO) Media Tubes
Pre-enrich food samples prior to the isolation of Salmonella species with ready prepared media tubes.
Brand: Thermo Scientific™ TV5013D
91.14 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Pre-enrich food samples prior to the isolation of Salmonella species with Thermo Scientific™ Buffered Peptone Water (ISO) in ready prepared media tubes. Buffered Peptone Water (ISO) is a non-selective pre-enrichment medium for the isolation of Salmonella species from food and associated samples.
Specifications
Ivory, transparent | |
Buffered Peptone Water | |
Germany | |
9 mL | |
50 tubes in a carton box |
2 to 25°C | |
Tube | |
Microbiology Media | |
9 ± 0.5mL | |
50/Pk. |
For In Vitro Diagnostic Use.