missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Buffered Zinc Formalin Histology Fixative
Enhance nuclear detail in immunohistochemistry techniques with Epredia™ Buffered Zinc Formalin.
£55.12 - £75.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10230289
|
Epredia™
5701ZF |
1 Gallon Bottle |
£75.00
Pack of 4 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10086258
|
Epredia™
5705ZF |
5 gallon Cubitainer |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Easy-to-pour bottle helps reduce spills.Specifications
Formalin |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P362-Take off contaminated clothing.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, methylalcohol, zinc sulphate, sodium acetate