Learn More
Thermo Scientific™ Buffers for GeneJET™ Blood RNA Kit
Purify high quality total RNA from fresh or frozen whole blood, as well as blood collected in PAXgene and Tempus Tubes
Brand: Thermo Scientific™ R2851
Description
The GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.
The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).
Highlights
•Versatile - compatible with fresh and frozen blood, buffy coat, as well as blood collected in PAXgene™and Tempus ™ Tubes
•Convenient - specially optimized Lysis Buffer is suitable for RNA stabilization up to 48 h
•Efficient - up to 4 μg of total RNA from 500 μL of healthy human blood
•Pure - isolates intact total RNA free of any RT-qPCR inhibitors
Applications
Isolated total RNA can be used in:
• RT-PCR
• RT-qPCR
• Northern Blotting
Includes
• Lysis Buffer
• Wash Buffer 1 (conc.)
• Wash Buffer 2 (conc.)
• DNase l (lyophilized)
• DNase I Reconstitution Buffer
• 2x DNase I Buffer
• Manganese Chloride Solution
• Water, nuclease-free
• GeneJET RNA Purification Columns pre-assembled with Collection Tubes
• Collection Tubes, 2 mL
• Collection Tubes, 1.5 mL
Hazardous:No
> Quality Control:Thermo Scientific GeneJET Stabilized and Fresh Whole Blood RNA Purification Mini Kit is qualified by isolating total RNA from 250 μL of human whole blood following the protocol outlined in the manual. The quality of isolated RNA is evaluated spectrophotometrically and by agarose gel electrophoresis. The purified RNA has an A260/A280 ratio between 1.9 and 2.1 and the RNA integrity number (RIN) of ≥ 8.
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Storage Condition:Ambient
Specifications
Store at room temperature | |
GeneJET Blood RNA Kit | |
24 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.