missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ Plasmid Kits
Additional supply of buffer solutions (Resuspension Buffer A1, Lysis Buffer A2, Neutralization Buffer A3, Wash Buffer AW, Wash Buffer A4 Concentrate, or Elution Buffer AE) for use in preparing high-purity plasmid DNA using a variety of NucleoSpin™ Plasmid kits.
Brand: Macherey-Nagel™ 740911.3
This item is not returnable.
View return policy
Description
For use with the following NucleoSpin™ Plasmid DNA purification kits:
- NucleoSpin™ Plasmid (Lid/NoLid)
- NucleoSpin™ Plasmid QuickPure
- NucleoSpin™ Plasmid EasyPure
- NucleoSpin™ 8/96 Plasmid (Full/Core Kit)
- NucleoSpin™ Plasmid Transfection-Grade
- NucleoSpin™ 96 Plasmid Transfection-Grade (Full/Core Kit)
Especificaciones
Colorless | |
7.5 to 8.5 | |
Odorless | |
Liquid | |
Resuspension Buffer A1 |
NucleoSpin™ Plasmid kits | |
3000 mL | |
Room Temperature (18°C to 25°C) | |
Preparation high-purity plasmid DNA |