missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin™ RNA Plus

Additional buffer solutions for use with NucleoSpin™ RNA Plus Kits, for efficient and quick RNA isolation from cells and tissues.

£33.35 - £75.25

Specifications

For Use With (Equipment) NucleoSpin RNA Plus kit
CAS 593-84-0
Odor Odorless
Content And Storage Room Temperature (18°C to 25°C)
Form Liquid
View More Specs

Products 2
Product Code Brand Product Type Quantity Price Quantity & Availability  
Product Code Brand Product Type Quantity Price Quantity & Availability  
15849127
View Documents
Macherey-Nagel™
740907.30
Binding Solution BS 30 mL
£33.35
30mL
Please to purchase this item. Need a web account? Register with us today!
15839127
View Documents
Macherey-Nagel™
740906.125
Lysis buffer LBP 125 mL
£75.25
125mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NucleoSpin™ RNA Plus Kits

  • gDNA Removal Columns included – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal using the same column (in one step)
  • Novel lysis buffer – exceptionally efficient, with no mercaptoethanol or TCEP necessary
  • Two column silica membrane system (one for DNA removal, one for RNA isolation)
  • Downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications

Specifications

NucleoSpin RNA Plus kit
Odorless
Liquid
593-84-0
Room Temperature (18°C to 25°C)
RNA isolation from cells and tissue
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : At least 1 year

SDS
Documents

Documents

Product Certifications