Learn More
Merck Millipore BugBuster™ 10X Protein Extraction Reagent
Reagent for protein sample preparation
Brand:  Merck Millipore 70921-4
483.33 GBP valid until 2024-12-20
Use promo code "21644" to get your promotional price.
Product Code. 10579953
  • £840.00 / 50mL
Edge
Explore more special offers
Description
Description
  • Concentrated formulation of proprietary detergents without addition of salts or buffer components
  • Provides flexible alternative to ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components
  • Consists all of the bioprocessing benefits of standard BugBuster plus the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein