Learn More
Fisherbrand™ Bulb Reflector
Illuminate Inverso TC100 Microscopes with this reflector. Fisherbrand™ Bulb Reflectors are ergonomically designed for everyday use.
Brand: Fisherbrand™ 7260.1211
This item is not returnable.
View return policy
Description
- Bulb Reflector
- Accessory for microscopes: FB69155 and FB69156
Specifications
Bulb Reflector | |
Episcopic light | |
Diamond |
12 V | |
2.5 to 120x |